Spex CAN-CAN-KIT Canadian Pesticide Kit 1, 100 µg/mL, 1 mL; 7 ampules/Kit

Representative image only.

Spex CAN-CAN-KIT Canadian Pesticide Kit 1, 100 µg/mL, 1 mL; 7 ampules/Kit

Spex | Item: 0455949

Analytical standards for medicinal and recreational cannabis testing helps ensure high-quality products

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 100 ppm Canadian pesticide kit in LC/MS acetonitrile:LC/MS methanol 50:50 matrix
  • Contains all 6 Canadian pesticide mixes (1, 2, 3, 4, 5, 6) and dibrom (naled) standard
  • Seven 1 mL ampules

More About this Item

While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants. Use this kit containing the 95 compounds that are required to be tested based on regulations set by the Government of Canada to test for the presence of unauthorized pesticides.

Spex CertiPrep® offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.

Note: Supplied in LC/MS Acetonitrile, except for mix 6 which is in LC/MS Acetonitrile:LC/MS Methanol 50:50.

Designed for state specific pesticide regulations:
• OAR 333-008-11
• HB 3460
• AOAC 2007-01
• EN 15662

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component kit contains Canadian Pesticide Mixes 1, 2, 3, 4, 5, and 6 and Dibrom (Naled).

• Canadian Pesticide Mix 1 is a 1 mL ampule which contains 100 µg/mL (100 ppm) of Aldicarb, Baygon (Propoxur), Carbaryl, Carbofuran, Fenoxycarb, Imazalil, Imidacloprid, Iprodione, Methiocarb, Methomyl, Novaluron, Oxamyl, Pirimicarb, and Thiacloprid. This is available individually as part number 04559-43.

• Canadian Pesticide Mix 2 is a 1 mL ampule which contains 100 µg/mL (100 ppm) of Allethrin, Baythroid (Cyfluthrin), Bifenthrin, Cypermethrin, Deltamethrin, Kinoprene, Methoprene, MGK 264™, Permethrin (mix of cis & trans), d-Phenothrin (mix of cis & trans), Piperonyl butoxide, Prallethrin (mix of isomers), Pyrethrins (mix of isomers), Resmethrin, Sanmarton, and Tetramethrin. This is available individually as part number 04559-44.

• Canadian Pesticide Mix 3 is a 1 mL ampule which contains 100 µg/mL (100 ppm) of Azadirachtin, Benzovindiflupyr, Endosulfan I, Endosulfan II, Endosulfan sulfate, Fipronil, Pentachloronitrobenzene, and Trifloxystrobin. This is available individually as part number 04559-45.

• Canadian Pesticide Mix 4 is a 1 mL ampule which contains 100 µg/mL (100 ppm) of Acephate, Chlorpyrifos, Coumaphos, Diazinon, Dichlorvos, Dimethoate, Etofenprox, Etoxazole, Fensulfothion, Fenthion, Imidan (Phosmet), Malathion, Methyl parathion, Mevinphos, Prophos, Spiroxamine, Terrazole, Tetrachlorvinphos (Z), and Thiophanate-methyl. This is available individually as part number 04559-46.

• Canadian Pesticide Mix 5 is a 1 mL ampule which contains 100 µg/mL (100 ppm) of Acequinocyl, Acetamiprid, Azoxystrobin, Bifenazate, Boscalid, Buprofezin, Chlorantraniliprole, Chlorfenapyr, Clofentezine, Clothianidin, Cyantraniliprole, Cyprodinil, Dinotefuran, (E)-Fenpyroximate, Flonicamid, Fludioxonil, Fluopyram, Folicur, Hexythiazox, Paclobutrazol, Propiconazole (Tilt), Pyraclostrobin, Systhane (Myclobutanil), Tebufenozide, Teflubenzuron, and Thiamethoxam. This is available individually as part number 04559-47.

• Canadian Pesticide Mix 6 is a 1 mL ampule which contains 100 µg/mL (100 ppm) of Abamectin, Daminozide, Dodemorph, Kresoxim-methyl, Metalaxyl, Pyridaben, Spinetoram, Spinosad, Spirodiclofen, Spiromesifen, and Spirotetramat. This is available individually as part number 04559-48.

• Dibromo (Naled) standard is a 1 mL ampule which contains 1,000 µg/mL (1,000 ppm) of dibromo (naled) standard. This is available individually as part number 04559-42.

Hazardous material shipping fees may apply.

Specifications
  • Concentration100 µg/mL (100 ppm)
  • MatrixMultiple
  • Element/IonAmpule 1: Canadian pesticide mix 1
    Ampule 2: Canadian pesticide mix 2
    Ampule 3: Canadian pesticide mix 3
    Ampule 4: Canadian pesticide mix 4
    Ampule 5: Canadian pesticide mix 5
    Ampule 6: Canadian pesticide mix 6
    Ampule 7: Dibromo (Naled) standard
  • FormLiquid
  • Volume (mL)1
  • Packaging7 x 1 mL ampules
  • DOT InformationHazardous; Acetonitrile; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Irritant
  • CAS NumberMultiple
  • UN NumberUN1648
  • Shelf Life (months)12
  • DescriptionCanadian Pesticide Kit, 100 µg/mL in LC/MS Acetonitrile, 1 mL; 7 ampules/Kit
Documents & Certificates
Reviews
Q&A