Spex CAN-CAN-KIT-2 Canadian Pesticide Kit 2, 1 mL; 6 ampules/Kit

Representative image only.

Spex CAN-CAN-KIT-2 Canadian Pesticide Kit 2, 1 mL; 6 ampules/Kit

Spex | Item: 0455956

Analytical standard for medicinal and recreational cannabis testing helps ensure high-quality products

£934.00GBP / Each
£1,120.80GBP (incl VAT)

£934.00GBP / Each
£1,120.80GBP (incl VAT)
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • Contains six Canadian pesticide mixes (1A, 2A, 4A, 5A, 6A, and 7) in LC/MS acetonitrile matrix
  • Six 1 mL ampules

More About this Item

While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.

Spex CertiPrep® offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.

Designed for state specific pesticide regulations:
• OAR 333-008-11
• HB 3460
• AOAC 2007-01
• EN 15663

For your convenience, we have designed cannabis pesticide mixes and a kit which contains the 95 compounds that are required to be tested based on regulations set by the Government of Canada. This requirement for mandatory testing for the presence of unauthorized pesticides will help ensure that Canadians can continue to have confidence in obtaining safe, quality-controlled medical cannabis from licensed producers.

This multi-component kit contains Canadian Pesticide Mixes 1A, 2A, 4A, 5A, 6A, and 7.

• Canadian Pesticide Mix 1A is a 1 mL ampule which contains 20 µg/mL (20 ppm) of Baygon (Propoxur), Carbofuran, Fenoxycarb, Imidacloprid, Methiocarb, Pirimicarb, Thiacloprid; 50 µg/mL (50 ppm) of Carbaryl, Imazalil, Methomyl, Novaluron; 1,000 µg/mL (1,000 ppm) of Aldicarb, Iprodione; and 3,000 µg/mL (3,000 ppm) of Oxamyl. This is available individually as part number 04559-50.

• Canadian Pesticide Mix 2A is a 1 mL ampule which contains 50 µg/mL (50 ppm) of MGK 264™, Phenothrin (cis & trans), Prallethrin, Pyrethrins (mix of isomers); 100 µg/mL (100 ppm) of Resmethrin, Sanmarton, Tetramethrin; 200 µg/mL (200 ppm) of Allethrin, Baythroid (Cyfluthrin), Piperonyl butoxide; 300 µg/mL (300 ppm) of Cypermethrin; 500 µg/mL (500 ppm) of Deltamethrin, Kinoprene; 1,000 µg/mL (1,000 ppm) of Permethrin; 1,000 µg/mL (1,000 ppm) of Bifenthrin; and 2,000 µg/mL (2,000 ppm) of Methoprene. This is available individually as part number 04559-51.

• Canadian Pesticide Mix 4A is a 1 mL ampule which contains 20 µg/mL (20 ppm) of Acephate, Coumaphos, Diazinon, Dimethoate, Etoxazole, Fensulfothion, Fenthion, Imidan (Phosmet), Malathion, Prophos, Tetrachlorvinphos (Z); 30 µg/mL (30 ppm) of Terrazole; 40 µg/mL (40 ppm) of Chlorpyrifos, Etofenprox, Methyl parathion, Mevinphos, Thiophanate-methyl; and100 µg/mL (100 ppm) of Dichlorvos, Spiroxamine. This is available individually as part number 04559-52.

• Canadian Pesticide Mix 5A is a 1 mL ampule which contains 20 µg/mL (20 ppm) of Azoxystrobin, Bifenazate, Boscalid, Buprofezin, Chlorantraniliprole, Clofentezine, Cyantraniliprole, E)-Fenpyroximate, Fludioxonil, Fluopyram, Paclobutrazol, Pyraclostrobin, Systhane (Myclobutanil), Tebufenozide, Thiamethoxam; 30 µg/mL (30 ppm) of Acequinocyl; 50 µg/mL (50 ppm) of Chlorfenapyr, Clothianidin, Flonicamid, Folicur, Teflubenzuron; 100 µg/mL (100 ppm) of Acetamiprid, Dinotefuran, Propiconazole (Tilt); and 250 µg/mL (250 ppm) of Cyprodinil. This is available individually as part number 04559-53.

• Canadian Pesticide Mix 6A is a 1 mL ampule which contains 20 µg/mL (20 ppm) of Kresoxim-methyl, Metalaxyl, Spinetoram, Spirotetramat; 50 µg/mL (50 ppm) of Dodemorph, Pyridaben; 100 µg/mL (100 ppm) of Abamectin, Daminozide, Spinosad; 250 µg/mL (250 ppm) of Spirodiclofen; and 3,000 µg/mL (3,000 ppm) of Spiromesifen. This is available individually as part number 04559-54.

• Canadian Pesticide Mix 7 is a 1 mL ampule which contains 20 µg/mL (20 ppm) of Benzovindiflupyr, Pentachloronitrobenzene, Trifloxystrobin; 50 µg/mL (50 ppm) of Dimethomorph, Endosulfan II, Endosulfan sulfate; 60 µg/mL (60 ppm) of Fipronil; 200 µg/mL (200 ppm) of Endosulfan I; and 1,000 µg/mL (1,000 ppm) of Azadirachtin. This is available individually as part number 04559-55.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixLC/MS Acetonitrile
  • Element/IonAmpule 1: Canadian pesticide mix 1AAmpule 2: Canadian pesticide mix 2AAmpule 3: Canadian pesticide mix 4AAmpule 4: Canadian pesticide mix 5AAmpule 5: Canadian pesticide mix 6AAmpule 6: Canadian pesticide mix 7
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Acetonitrile; Class 3, Packing Group II
  • Storage SymbolFreezer
  • UN NumberUN1648
  • CAS NumberSee individual mix
  • Shelf Life (months)12
  • DescriptionCanadian Pesticide Kit 2, Mixes 1A, 2A, 4A, 5A, 6A, and 7 in LC/MS Acetonitrile, 1 mL; 6 ampules/Kit
Documents & Certificates
Reviews
Q&A