Spex CAN-CAN-5A Canadian Pesticide Mix 5A in LC/MS Acetonitrile; 1 mL

Representative image only.

Spex CAN-CAN-5A Canadian Pesticide Mix 5A in LC/MS Acetonitrile; 1 mL

Spex | Item: 0455953

Analytical standard for medicinal and recreational cannabis testing helps ensure high-quality products

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • Multi-component Canadian pesticide mix 5A in LC/MS acetonitrile matrix
  • 1 mL ampule

More About this Item

While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.

Spex CertiPrep® offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.

Designed for state specific pesticide regulations:
• OAR 333-008-11
• HB 3460
• AOAC 2007-01
• EN 15663

For your convenience, we have designed cannabis pesticide mixes and a kit which contains the 95 compounds that are required to be tested based on regulations set by the Government of Canada. This requirement for mandatory testing for the presence of unauthorized pesticides will help ensure that Canadians can continue to have confidence in obtaining safe, quality-controlled medical cannabis from licensed producers.

This multi-component standard contains 26 components.

• Canadian Pesticide Mix 5A is a 1 mL ampule which contains 10 µg/mL (10 ppm) of Hexythiazox; 20 µg/mL (20 ppm) of Azoxystrobin, Bifenazate, Boscalid, Buprofezin, Chlorantraniliprole, Clofentezine, Cyantraniliprole, E)-Fenpyroximate, Fludioxonil, Fluopyram, Paclobutrazol, Pyraclostrobin, Systhane (Myclobutanil), Tebufenozide, Thiamethoxam; 30 µg/mL (30 ppm) of Acequinocyl; 50 µg/mL (50 ppm) of Chlorfenapyr, Clothianidin, Flonicamid, Folicur, Teflubenzuron; 100 µg/mL (100 ppm) of Acetamiprid, Dinotefuran, Propiconazole (Tilt); and 250 µg/mL (250 ppm) of Cyprodinil.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixLC/MS Acetonitrile
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Acetonitrile; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Irritant
  • CAS Number78587-05-0, 131860-33-8, 149877-41-8, 188425-85-6, 69327-76-0, 500008-45-7, 74115-24-5, 736994-63-1, 134098-61-6, 131341-86-1, 658066-35-4, 76738-62-0, 175013-18-0, 88671-89-0, 112410-23-8, 153719-23-4, 57960-19-7, 122453-73-0, 210880-92-5, 158062-67-0, 107534-96-3, 83121-18-0, 135410-02-7, 165252-70-0, 60207-90-1, 121552-61-2
  • UN NumberUN1648
  • Shelf Life (months)12
  • DescriptionCanadian Pesticide Mix 5A in LC/MS Acetonitrile; 1 mL
Documents & Certificates
Reviews
Q&A