Spex CALMIX4-500 Assurance® Grade Instrument Check Standard 4 for AA & ICP; 500 mL

Representative image only.

Spex CALMIX4-500 Assurance® Grade Instrument Check Standard 4 for AA & ICP; 500 mL

Spex | Item: 0456789

Multi-element instument check standards for AA and ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Calibrate and verify wavelength accuracy and stability in sequential and simultaneous ICP units
  • Instrument check standard 4 for AA & ICP in 2% HNO3 matrix
  • 500 mL bottle

More About this Item

Each Instrument Check Standard is designed to give the user wavelength ranges from 160 to 790 nm. Every ICP manufacturer has a specific group of elements at varying concentrations to determine instrument accuracy and reliability. Some have special calibration programs incorporated into their software; others give you information in their manuals.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 12 elements.

• Instrument Check Standard 4 is a 500 mL bottle which contains 50 µg/mL (50 ppm) of Potassium; 10 µg/mL (10 ppm) of Aluminum, Arsenic, Copper, Lead, Manganese, Nickel, Phosphorus, Scandium, Sodium, Zinc; and 1 µg/mL (1 ppm) of Barium.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeAssurance® Grade
  • ConcentrationMultiple
  • Matrix2% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Instrument Check Standard 4 for AA & ICP in 2% HNO3; 500 mL
Documents & Certificates
Reviews
Q&A