Spex C1001-09.4 Method C1001-09.4 Phthalates, 1,000 µg/mL in Isooctane; 1 mL

Representative image only.

Spex C1001-09.4 Method C1001-09.4 Phthalates, 1,000 µg/mL in Isooctane; 1 mL

Spex | Item: 0456220

Consumer safety standard helps meet guidelines set by agencies to protect consumers

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm PVC phthalate standard in isooctane matrix

More About this Item

Polyvinyl chloride, or PVC, is a very common plastic used in a wide range of common consumer products from children’s toys and care items to building and construction materials. In the US, ASTM (American Society for Testing and Materials) and CPSC (Consumer Product Safety Commission) have designed methods for testing children’s toys and childcare articles for compliance with the restriction of use for eight designated phthalates.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 8 components.

• Method C1001-09.4 Phthalates is a 1 mL ampule which contains 1,000 µg/mL (1,000 ppm) of Bis(2-Ethylhexyl)phthalate, Butylbenzyl phthalate, Di-n-butyl phthalate, Di-n-hexyl phthalate, Diamyl phthalate, Dicyclohexyl phthalate, Diisobutyl phthalate, and Diisononyl phthalate.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixIsooctane
  • Element/IonBis(2-Ethylhexyl)phthalate, Butylbenzyl phthalate, Di-n-butyl phthalate, Di-n-hexyl phthalate, Diamyl phthalate, Dicyclohexyl phthalate, Diisobutyl phthalate, Diisononyl phthalate
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Octanes; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1262
  • CAS Number117-81-7, 85-68-7, 84-74-2, 84-75-3, 131-18-0, 84-61-7, 84-69-5, 28553-12-0
  • Shelf Life (months)12
  • DescriptionMethod C1001-09.4 Phthalates, 1,000 µg/mL in Isooctane; 1 mL
Documents & Certificates
Reviews
Q&A