Spex BFM-10Y Biodiesel Standard, 5 Elements, 10 µg/g in B100 Biofuel; 100 mL

Representative image only.

Spex BFM-10Y Biodiesel Standard, 5 Elements, 10 µg/g in B100 Biofuel; 100 mL

Spex | Item: 0457028

Standards specifically designed for industrial use

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Manufactured under an internationally accredited ISO 9001 quality system and compliant with ISO/IEC 17025
  • Meet the requirements for testing ASTM Methods D6751, D5453 and EN 14216
  • 10 ppm multi-element biodiesel standard in B100 biofuel matrix
  • 100 mL bottle

More About this Item

Governments worldwide have passed regulations that mandate lower levels of sulfur in biodiesel fuel. To comply with the implementation of these regulations, SPEX CertiPrep offers specifically designed Certified Reference Materials for industrial use. Our B100 Biodiesel Standards meet the requirements for testing ASTM Methods D6751, D5453 and EN 14214.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration10 µg/g (10 ppm)
  • MatrixB100 Biofuel
  • Element/IonCalcium, Magnesium, Phosphorus, Potassium, and Sodium
  • FormLiquid
  • Volume (mL)100
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionBiodiesel Standard, 5 Elements, 10 µg/g in B100 Biofuel; 100 mL
Documents & Certificates
Reviews
Q&A