Spex BD-3515 Tricaprin Glyceride Impurity Standard, 8,000 µg/mL; 5 mL

Representative image only.

Spex BD-3515 Tricaprin Glyceride Impurity Standard, 8,000 µg/mL; 5 mL

Spex | Item: 0456143

Biodiesel standard designed for ASTM D6584 & EN14105

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA ASTM Method D6584 and EN14105
  • 8,000 ppm biodiesel fuel standard in pyridine matrix
  • 5 mL bottle

More About this Item

Biodiesel is fast becoming an environmentally friendly alternative to petroleum sources. This fuel source is obtained by esterification of oils derived from plants or animals. To meet the demands of this industry, Cole-Parmer offers Biodiesel Certified Reference Materials, as well as glyceride and FAME (Fatty Acids Methyl Esters) impurity standards; all designed to save time and money by eliminating the need to prepare in-house standards.

Our standards are manufactured from the highest purity starting materials and the highest grade of solvents available to guarantee superior quality. These standards are manufactured in a specially designed laboratory built to eliminate contamination issues that may arise during the manufacturing process. The standards then go through a rigorous QC process where a senior chemist approves each lot. This guarantees the standards are accurate and stable at the stated concentrations for all of the components. In addition, all standards are supplied with a detailed, comprehensive Certificate of Analysis.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration8,000 µg/mL (8,000 ppm)
  • MatrixPyridine
  • Element/IonTricaprin
  • FormLiquid
  • Volume (mL)5
  • DOT InformationHazardous; Pyridine; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Irritant
  • UN NumberUN1282
  • CAS Number621-71-6
  • Shelf Life (months)36
  • DescriptionTricaprin Glyceride Impurity Standard, 8,000 µg/mL in Pyridine; 5 mL
Documents & Certificates
Reviews
Q&A