Spex BASE75-G Base Oil 75 Blank; 1 Gallon

Representative image only.

Spex BASE75-G Base Oil 75 Blank; 1 Gallon

Spex | Item: 0457015

Same certified base oils used in our single and multi-element blends

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • 1 gallon bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Specifications
  • MatrixBase Oil 75
  • FormLiquid
  • Volume (mL)3785
  • DOT InformationNon-Hazardous:: Not regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionBase Oil 75 Blank; 1 Gallon
Documents & Certificates
Reviews
Q&A