Spex BASE20-G Base Oil 20 Blank, 1 Gallon
Representative image only.
Spex BASE20-G Base Oil 20 Blank, 1 Gallon
Spex | Item: 0457013
Certified base oil used in single and multi-element blends
Call For Price
Call For Price
Product Description
Includes
Supplier Certificate of Analysis (CoA) is included with product shipment.
Key Features
  • Determine wear metals in engine oils and lubricants
  • ISO 9001 and ISO/IEC 17025 compliant
More About this Item
The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc.
By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Specifications
  • ClassOrganometallic Standards
  • Element/IonBase Oil 20
  • Volume (mL)3785
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionBase Oil 20 Blank, 1 Gallon