Spex AS-SO4S9-2Y Sulfate-Sulfur Single-Element Ion Anion Standard, 1,000 µg/mL (1,000 ppm) in H2O, 125 mL

Representative image only.

Spex AS-SO4S9-2Y Sulfate-Sulfur Single-Element Ion Anion Standard, 1,000 µg/mL (1,000 ppm) in H2O, 125 mL

Spex | Item: 0450151

Single and multi-element standards for ion chromatography

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Traceable standards
  • Perfect for the analytical laboratory

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. To ensure accurate analysis, quality standards which are traceable and stable are necessary. Spex CertiPrep®offers the highest quality IC standards available for the analytical laboratory.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixWater
  • Element/IonSulfate-Sulfur
  • FormLiquid
  • Volume (mL)125
  • Storage SymbolAmbient
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionSulfate-Sulfur Single-Element Ion Anion Standard, 1,000 µg/mL (1,000 ppm) in H2O, 125 mL
Documents & Certificates
Reviews
Q&A