Supplier Certificate of Analysis (CoA) is included with product shipment.
Key Features
Ideal for calibrating and performing QA/QC checks on ion chromatography systems
Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
Traceable standards ensure accurate analysis
Fluoride ISE standard set contains 5 solutions
10 ppm, 100 ppm, 1000 ppm, and 0.1 M fluoride (F-) ion selective standards and low level TISAB II buffer in H2O matrix
More About this Item
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers the highest quality IC standards available for the analytical laboratory.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
This multi-element standard set contains 5 solutions.
• Solution 1 is a 125 mL bottle which contains 0.1 M fluoride (F-) ion in H2O.
• Solution 2 is a 125 mL bottle which contains 10 µg/mL (10 ppm) fluoride (F-) ion in H2O.
• Solution 3 is a 125 mL bottle which contains 100 µg/mL (100 ppm) fluoride (F-) in H2O.
• Solution 4 is a 125 mL bottle which contains 1,000 µg/mL (1,000 ppm) fluoride (F-) in H2O.
• Solution 5 is a 500 mL bottle which contains low level TISAB II buffer in H2O.
Hazardous material shipping fees may apply.
Specifications
ConcentrationMultiple
MatrixH2O
Element/IonFive solution set
FormLiquid
DOT InformationNot regulated
Storage SymbolAmbient
Hazard SymbolNot Regulated
UN NumberNot regulated
Shelf Life (months)12
DescriptionFluoride (F-) Ion Selective Electrode Standard Set, 5 Solutions