Spex AS-F9-1AY Fluoride (F-) Ion Selective Electrode Standard, 10 µg/mL; 125 mL

Representative image only.

Spex AS-F9-1AY Fluoride (F-) Ion Selective Electrode Standard, 10 µg/mL; 125 mL

Spex | Item: 0450137

Single-element ion selective electrode standards for ion chromatography

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Ideal for calibrating and performing QA/QC checks on ion chromatography systems
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Traceable standards ensure accurate analysis
  • 10 ppm fluoride ion selective electrode standard in H2O matrix
  • 125 mL bottle

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers the highest quality IC standards available for the analytical laboratory.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration10 µg/mL (10 ppm)
  • MatrixH2O
  • Element/IonFluoride (F-)
  • FormLiquid
  • Volume (mL)125
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionFluoride (F-) Single-Element Ion Selective Electrode Standard, 10 µg/mL in H2O; 125 mL
Documents & Certificates
Reviews
Q&A