Spex AS-CL9-1X Chloride Single-Element Ion Anion Standard, 100 µg/mL in H2O; 500 mL

Representative image only.

Spex AS-CL9-1X Chloride Single-Element Ion Anion Standard, 100 µg/mL in H2O; 500 mL

Spex | Item: 0450164

Single element standard for ion chromatography

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Ideal for a variety of different kinds of charged analytes
  • Manufactured under ISO ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Contains 100 ppm of Chloride (CI)- in H20 matrix

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary.

These solutions offer the highest quality IC standards available for the analytical laboratory. Spex CertiPrep®Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Specifications
  • Concentration100 µg/mL (100 ppm)
  • MatrixH2O
  • ClassIC
  • Element/IonChloride
  • FormLiquid
  • Volume (mL)500
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot regulated
  • UN NumberNot regulated
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionChloride Single-Element Ion Anion Standard, 100 µg/mL in H2O; 500 mL
Documents & Certificates
Reviews
Q&A