Spex AM-SET Assurance® Grade Alternate Metals Standard Set; 125 mL

Representative image only.

Spex AM-SET Assurance® Grade Alternate Metals Standard Set; 125 mL

Spex | Item: 0456855

Multi-element groundwater and wastewater pollution control check standards for AA & ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Refer to US EPA Methods Manual 600/4-79-020 "Methods for Chemical Analysis of Water and Wastes" Trace Metals 21I, 21II and 21III Methods
  • Alternate metals standard set contains 2 solutions
  • Two 125 mL bottles

More About this Item

May be used either as standards or as a means to check the individual analysts accuracy and precision.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This alternate metals standard set contains 2 solutions.

• Solution 1 is a 125 mL bottle which contains 20 µg/mL (20 ppm) of Aluminum, Iron, Vanadium; 10 µg/mL (10 ppm) of Cobalt, Copper, Manganese, Nickel, Zinc; and 5 µg/mL (5 ppm) of Antimony, Beryllium, Thallium in 2% HNO3. This is available individually as part number 04568-49.

• Solution 2 is a 125 mL bottle which contains 500 µg/mL (500 ppm) of Calcium, Sodium and 100 µg/mL (100 ppm) of Magnesium, Potassium in 2% HNO3. This is available individually as part number 04568-51.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeAssurance® Grade
  • ConcentrationMultiple
  • Matrix2% HNO3
  • Element/IonTwo bottle set
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Grup III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionSpex CertiPrep Assurance® Grade Alternate Metals Standard Set for AA & ICP in 2% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A