Spex 8310-A PAH Combination Mix for US EPA Method 8310, 500 µg/mL; 1 mL

Representative image only.

Spex 8310-A PAH Combination Mix for US EPA Method 8310, 500 µg/mL; 1 mL

Spex | Item: 0455728

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8310
  • 500 ppm polynuclear aromatic hydrocarbon (PAH) combination mix in acetonitrile matrix
  • 1 mL ampule

More About this Item

US EPA Method 8310 is an analytical method for the monitoring of PNA hydrocarbons in groundwater and waste samples by HPLC using UV and fluorescence detectors.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 16 components.

• Polynuclear Aromatic Hydrocarbon (PAH) Combination Mix is a 1 mL ampule which contains 500 µg/mL (500 ppm) of Acenaphthene, Acenaphthylene, Anthracene, Benz(a)anthracene, Benzo(a)pyrene, Benzo(b)fluoranthene, Benzo(g,h,i)perylene, Benzo(k)fluoranthene, Chrysene, Dibenz(a,h)anthracene, Fluoranthene, Fluorene, Indeno(1,2,3-cd)pyrene, Naphthalene, Phenanthrene, and Pyrene.

Hazardous material shipping fees may apply.

Specifications
  • Concentration500 µg/mL (500 ppm)
  • MatrixAcetonitrile
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Acetonitrile; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • CAS Number83-32-9, 208-96-8, 120-12-7, 56-55-3, 50-32-8, 205-99-2, 191-24-2, 207-08-9, 218-01-9, 53-70-3, 206-44-0, 86-73-7, 193-39-5, 91-20-3, 85-01-8, 129-00-0
  • UN NumberUN1648
  • Shelf Life (months)36
  • DescriptionPolynuclear Aromatic Hydrocarbon (PAH) Combination Mix, 500 µg/mL in Acetonitrile; 1 mL
Documents & Certificates
Reviews
Q&A