Spex 8270-IXB-PK Appendix IX Mix B for US EPA Method 8270, 2,000 µg/mL; 1 mL; 4/pk

Representative image only.

Spex 8270-IXB-PK Appendix IX Mix B for US EPA Method 8270, 2,000 µg/mL; 1 mL; 4/pk

Spex | Item: 0500914

Analytical standards for the monitoring of semivolatiles in environmental and waste samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8270
  • 2,000 ppm appendix IX mix B in methylene chloride:acetone:benzene matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethyl Tertiary-Butyl Ether
  • ChemistryOrganics;Standards
  • Element/IonTrichloroacetic acid
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methyl tert-butyl ether; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN2398
  • CAS Number76-03-9
  • Shelf Life (months)36
  • DescriptionTrichloroacetic Acid Organic Standard, 1,000 µg/mL in Methyl Tertiary-Butyl Ether; 1 mL
Documents & Certificates
Reviews
Q&A