Spex 8270-AF-C Nitrosamines Mix for US EPA Method 8270, 2,000 µg/mL; 1 mL

Representative image only.

Spex 8270-AF-C Nitrosamines Mix for US EPA Method 8270, 2,000 µg/mL; 1 mL

Spex | Item: 0455720

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8270
  • 2,000 ppm nitrosamines mix in methylene chloride matrix
  • 1 mL ampule

More About this Item

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 10 components.

• Nitrosamines Mix is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of n-Nitrosodi-n-butylamine, n-Nitrosodiethylamine, n-Nitrosodimethylamine, n-Nitrosodiphenylamine, n-Nitrosodi-n-propylamine, n-Nitrosomethylethylamine, n-Nitrosomorpholine, n-Nitrosopiperidine, and n-Nitrosopyrrolidine.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethylene Chloride
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Dichloromethane; Class 6.1, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • CAS Number924-16-3, 55-18-5, 62-75-9, 86-30-6, 621-64-7, 10595-95-6, 59-89-2, 100-75-4, 930-55-2
  • UN NumberUN1593
  • Shelf Life (months)24
  • DescriptionNitrosamines Mix, 2,000 µg/mL in Methylene Chloride; 1 mL
Documents & Certificates
Reviews
Q&A