Spex 8100-A Polynuclear Aromatic Hydrocarbon (PAH) Mix A, 2,000 µg/mL; 1 mL

Representative image only.

Spex 8100-A Polynuclear Aromatic Hydrocarbon (PAH) Mix A, 2,000 µg/mL; 1 mL

Spex | Item: 0456596

Semivolatile organic standards for testing groundwater, soil, sludge, and sediment

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8100
  • 2,000 ppm PAH mix A in methylene chloride:benzene matrix
  • 1 mL ampule

More About this Item

US EPA Method 8100 is an analytical method for the monitoring of PNA hydrocarbons in aqueous and solid samples by GC/FID.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 17 components.

• Polynuclear Aromatic Hydrocarbon (PAH) Mix A is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of Acenaphthene, Acenaphthylene, Anthracene, Benz(a)anthracene, Benzo(a)pyrene, Benzo(b)fluoranthene, Benzo(g,h,i)perylene, Benzo(k)fluoranthene, Carbazole, Chrysene, Dibenz(a,h)anthracene, Fluoranthene, Fluorene, Indeno(1,2,3-c,d)pyrene, Naphthalene, Phenanthrene, and Pyrene.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethylene Chloride:Benzene
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Flammable liquids, toxic, n.o.s. (Benzene, Dichloromethane); Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Irritant, Health Hazard
  • UN NumberUN1992
  • CAS Number83-32-9, 208-96-8, 120-12-7, 56-55-3, 50-32-8, 205-99-2, 191-24-2, 207-08-9, 86-74-8, 218-01-9, 53-70-3, 206-44-0, 86-73-7, 193-39-5, 91-20-3, 85-01-8, 129-00-0
  • Shelf Life (months)36
  • DescriptionPolynuclear Aromatic Hydrocarbon (PAH) Mix A, 2,000 µg/mL (2,000 ppm) in Methylene Chloride:Benzene; 1 mL
Documents & Certificates
Reviews
Q&A