Spex 8082-IC Initial Combination Calibration Standard, 1,000 µg/mL; 1 mL

Representative image only.

Spex 8082-IC Initial Combination Calibration Standard, 1,000 µg/mL; 1 mL

Spex | Item: 0455896

Test for pescides and PBCs in soil, source water, drinking water, and waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8082 and CLP Series Methods
  • 1,000 ppm initial combination calibration standard in hexane matrix
  • 1 mL ampule

More About this Item

The EPA is tasked with the monitoring of environmental systems as they pertain to contamination and human health. The methods issued by the EPA are created to respond to specific toxins or persistent organic pollutants (POPs) found in environmental samples such as soil, source water, drinking water, and waste. In particular, pollutants such as polychlorinated biphenyls (PCBs), which were in widespread industrial use up until their restriction, are of concern due to their stable and persistent nature in the environment. Another group of chemicals of high concern is the hundreds of commercial pesticides and herbicides in use in the world today. Pesticides, from algaecides to virucides, are used in large quantities in industrial and private agriculture. The concern over human pesticide exposure over the past few decades has led to extensive monitoring of these pesticides. It has been reported that over 98% of insecticides and 95% of herbicides affect areas other than their intended target product. It is essential that monitoring agencies have accurate standard mixes to measure the pesticide levels in the environment.

Many new pesticides are now being tested using highly sensitive LC/MS techniques, in addition to traditional GC techniques, to determine minute amounts of residue in environmental samples and food products. We facilitate ease of monitoring and testing of pesticides by creating pesticide test mixes to suit your monitoring needs. Our standards are designed to work within EPA, AOAC and FDA analytical testing methods using all of the leading analytical techniques: LC, LC/MS, GC, and GC/MS. In addition, custom pesticide mixes can be made to your specifications to create a mix that meets your needs.

Applicable Methods:
• US EPA Method 8082 is an analytical method for the monitoring of PCBs in environmental samples by GC/ECD or GC/ELCD
• CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Pesticides Analysis Method is an analytical method for the analysis of aqueous or solid samples for pesticides by GC/ECD.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component solution contains 2 components.

• Initial Combination Calibration Standard is a 1 mL ampule which contains 1,000 µg/mL (1,000 ppm) of Aroclor 1016 and Aroclor 1260.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixHexane
  • Element/IonAroclor 1016, Aroclor 1260
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Hexanes; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • UN NumberUN1208
  • CAS Number12674-11-2, 11096-82-5
  • Shelf Life (months)36
  • DescriptionInitial Combination Calibration Standard, 1,000 µg/mL in Hexane; 1 mL
Documents & Certificates
Reviews
Q&A