Spex 8061-X Phthalate Esters Semivolatiles Mix, 2,000 µg/mL in Hexane; 1 mL

Representative image only.

Spex 8061-X Phthalate Esters Semivolatiles Mix, 2,000 µg/mL in Hexane; 1 mL

Spex | Item: 0456595

Semivolatile organic standards for testing groundwater, soil, sludge, and sediment

£118.00GBP / Each
£141.60GBP (incl VAT)

£118.00GBP / Each
£141.60GBP (incl VAT)
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8061
  • 2,000 ppm phthalate esters semivolatiles mix in hexane matrix
  • 1 mL ampule

More About this Item

US EPA Method 8061 is an analytical method for the monitoring of phthalate esters in aqueous and solid samples by GC/MS or GC/ECD.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 15 components.

• Phthalate Esters Semivolatiles Mix is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of Bis(2-Ethylhexyl)phthalate, Bis(2-methoxyethyl)phthalate, Bis(2-n-butoxyethyl)phthalate, Bis(4-methyl-2-pentyl)phthalate, Butylbenzyl phthalate, Diamyl phthalate, Dicyclohexyl phthalate, Diethyl phthalate, Diisobutyl phthalate, Dimethyl phthalate, Di-n-butyl phthalate, Di-n-hexyl phthalate, Di-n-octyl phthalate, Dinonyl phthalate, and Hexyl 2-ethylhexyl phthalate.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixHexane
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Hexanes; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1208
  • CAS Number117-81-7, 117-82-8, 117-83-9, 146-50-9, 85-68-7, 131-18-0, 84-61-7, 84-66-2, 84-69-5, 131-11-3, 84-74-2, 84-75-3, 117-84-0, 84-76-4, 75673-16-4
  • Shelf Life (months)36
  • DescriptionPhthalate Esters Semivolatiles Mix, 2,000 µg/mL in Hexane; 1 mL
Documents & Certificates
Reviews
Q&A