Spex 8020-A-PK Mix A for GC/PID for US EPA Method 8021, 2,000 µg/mL; 1 mL; 4/pk

Representative image only.

Spex 8020-A-PK Mix A for GC/PID for US EPA Method 8021, 2,000 µg/mL; 1 mL; 4/pk

Spex | Item: 0500897

Analytical standards for monitoring VOCs in a variety of solid waste matrices

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8021
  • 2,000 ppm mix A for GC/PID in methanol-P&T matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 8021 is a general analytical method for the monitoring of VOCs by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryOrganics;Standards
  • Element/Ion2,6-Diaminotoluene
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packin Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Irritant, Health Hazard
  • CAS Number823-40-5
  • UN NumberUN1230
  • Shelf Life (months)36
  • Description2,6-Diaminotoluene Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A