Spex 8015B-A Alcohols Mix for US EPA Method 8015, 2,000 µg/mL; 1 mL

Representative image only.

Spex 8015B-A Alcohols Mix for US EPA Method 8015, 2,000 µg/mL; 1 mL

Spex | Item: 0455662

Volatile organic standards for the monitoring of certain nonhalogenated VOCs and SVOCs

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8015
  • 2,000 ppm alcohol mix in DI water matrix
  • 1 mL ampule

More About this Item

US EPA Method 8015 is a general analytical method for the monitoring of certain non-halogenated VOCs and SVOCs by GC/FID.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 9 components.

• Alcohols Mix is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of Allyl alcohol, 1-Butanol, tert-Butyl alcohol, Ethanol, Ethylene glycol, Methanol, 2-Methyl-1-propanol, 1-Propanol, and 2-Propanol.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixDI Water
  • Element/IonAllyl alcohol, 1-Butanol, tert-Butyl alcohol, Ethanol, Ethylene glycol, Methanol, 2-Methyl-1-propanol, 1-Propanol, 2-Propanol
  • FormLiquid
  • Volume (mL)1
  • DOT InformationNot Regulated
  • Storage SymbolRefrigerator
  • Hazard SymbolHealth Hazard
  • UN NumberNot Regulated
  • CAS Number107-18-6, 71-36-3, 75-65-0, 64-17-5, 107-21-1, 67-56-1, 78-83-1, 71-23-8, 67-63-0
  • Shelf Life (months)36
  • DescriptionAlcohols Mix, 2,000 µg/mL in DI Water; 1 mL
Documents & Certificates
Reviews
Q&A