Spex 8015-OX-PK 8015-OX Mix for US EPA Method 8015, 2,000 µg/mL; 1 mL; 4/pk

Representative image only.

Spex 8015-OX-PK 8015-OX Mix for US EPA Method 8015, 2,000 µg/mL; 1 mL; 4/pk

Spex | Item: 0500894

Analytical standards for monitoring non-halogenated VOCs and SVOCs by GC/FID

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8015
  • 2,000 ppm 8015-OX mix in methanol-P&T matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 8015 is a general analytical method for the monitoring of certain non-halogenated VOCs and SVOCs by GC/FID.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryOrganics;Standards
  • Element/IonDyfonate (Fonofos)
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number944-22-9
  • UN NumberUN1230
  • Shelf Life (months)36
  • DescriptionDyfonate (Fonofos) Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A