Spex 625-PH Organochlorine Pesticides Mix, 2,000 µg/mL in Benzene; 1 mL

Representative image only.

Spex 625-PH Organochlorine Pesticides Mix, 2,000 µg/mL in Benzene; 1 mL

Spex | Item: 0455714

Analytical standards for monitoring of municipal, industrial wastewater, and waste samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 625 and 8270
  • 2,000 ppm organochlorine pesticides mix in benzene matrix
  • 1 mL ampule

More About this Item

US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixBenzene
  • Element/IonAldrin; alpha-BHC; beta-BHC; delta-BHC; Dieldrin; Endosulfan I; Endosulfan II; Endosulfan sulfate; Endrin; Endrin aldehyde; Endrin ketone; gamma-BHC; Heptachlor; Heptachlor epoxide (Isomer B); Methoxychlor; p,p’-DDD; p,p’-DDE; p,p’-DDT
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Benzene; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Irritant, Health Hazard
  • CAS Number309-00-2, 319-84-6, 319-85-7, 319-86-8, 60-57-1, 959-98-8, 33213-65-9, 1031-07-8, 72-20-8, 7421-93-4, 53494-70-5, 58-89-9, 76-44-8, 1024-57-3, 72-43-5, 72-54-8, 72-55-9, 50-29-3
  • UN NumberUN1114
  • Shelf Life (months)36
  • DescriptionOrganochlorine Pesticides Mix for US EPA Methods 625 and 8270, 2,000 µg/mL in Benzene; 1 mL
Documents & Certificates
Reviews
Q&A