Spex 605-X-PK Benzidines Mix for EPA Method 605, 8270 & CLP Series Methods, 2,000 µg/mL; 1 mL; 4/pk

Representative image only.

Spex 605-X-PK Benzidines Mix for EPA Method 605, 8270 & CLP Series Methods, 2,000 µg/mL; 1 mL; 4/pk

Spex | Item: 0500911

Analytical standard for the analysis of industrial and environmental wastewater

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 2,000 ppm benzidine mix in methanol matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 605 is an analytical method for the monitoring of benzidine in municipal and industrial wastewater by HPLC and an Electrochemical Detector.

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

CLP Series - The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Semivolatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for SVOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryOrganics;Standards
  • Element/IonChlorobenzene-d5, 1,4-Dichlorobenzene-d4, 1,4-Difluorobenzene, Pentafluorobenzene
  • FormLiquid
  • Volume (mL)15
  • Packaging4 x 15 mL
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • UN NumberUN1230
  • CAS Number3114-55-4, 3855-82-1, 540-36-3, 363-72-4
Documents & Certificates
Reviews
Q&A