Spex 5522-A-PK Free Acids Analyte Mix (10 Compts) for US EPA Method 552; 1 mL; 4/pk

Representative image only.

Spex 5522-A-PK Free Acids Analyte Mix (10 Compts) for US EPA Method 552; 1 mL; 4/pk

Spex | Item: 0500910

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 552
  • Free acid analyte mix (10 components) in methyl tertiary-butyl ether matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 552 is an analytical method for the monitoring of haloacetic acids and phenolic disinfectant by-products in drinking water by GC/ECD.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 10 components.

• Free Acids Analyte Mix is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of Tribromoacetic acid; 1,000 µg/mL (1,000 ppm) of Chlorodibromoacetic acid; 600 µg/mL (600 ppm) of Chloroacetic acid and Dichloroacetic acid; 400 µg/mL (400 ppm) of Bromoacetic acid, Bromochloroacetic acid, Bromodichloroacetic acid, and Dalapon; and 200 µg/mL (200 ppm) of Dibromoacetic acid and Trichloroacetic acid.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol
  • ChemistryOrganics;Standards
  • Element/Ionp,p'-DDT
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number50-29-3
  • UN NumberUN1230
  • Shelf Life (months)36
  • Descriptionp,p'-DDT Organic Standard, 1,000 µg/mL in Methanol; 1 mL
Documents & Certificates
Reviews
Q&A