Spex 5252-PCS-PK Performance Check Solution for US EPA Method 525, 500 µg/mL; 1 mL; 4/pk

Representative image only.

Spex 5252-PCS-PK Performance Check Solution for US EPA Method 525, 500 µg/mL; 1 mL; 4/pk

Spex | Item: 0500908

Analytical standards for the monitoring of semivolatiles in drinking water and raw water sources

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 525
  • 500 ppm performance check solution in methylene chloride matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 525 is an analytical method for the monitoring of semivolatiles in drinking water and raw water sources by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryStandards;Organics
  • Element/IonEpichlorohydrin
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Irritant, Health Hazard
  • UN NumberUN1230
  • CAS Number106-89-8
  • Shelf Life (months)24
  • DescriptionEpichlorohydrin Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A