Spex 5252-PA Organochlorine Pesticides Mix A, 200 µg/mL in Acetone; 1 mL

Representative image only.

Spex 5252-PA Organochlorine Pesticides Mix A, 200 µg/mL in Acetone; 1 mL

Spex | Item: 0455921

Analytical standards for medicinal and recreational cannabis testing helps ensure high-quality products

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 200 ppm organochlorine pesticide mix A in acetone matrix
  • 1 mL ampule

More About this Item

While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.

Spex CertiPrep offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.

Designed for state specific pesticide regulations:
• OAR 333-008-11
• HB 3460
• AOAC 2007-01
• EN 15662

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 18 components.

• Organochlorine Pesticides Mix A is a 1 mL ampule which contains 200 µg/mL (200 ppm) of Aldrin, alpha-BHC, beta-BHC, delta-BHC, Dieldrin, Endosulfan I, Endosulfan II, Endosulfan sulfate, Endrin, Endrin aldehyde, Endrin ketone, gamma-BHC, Heptachlor, Heptachlor epoxide (Isomer B), Methoxychlor, p,p’-DDD, p,p’-DDE, and p,p’-DDT.

Hazardous material shipping fees may apply.

Specifications
  • Concentration200 µg/mL (200 ppm)
  • MatrixAcetone
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Acetone; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant
  • CAS Number309-00-2, 319-84-6, 319-85-7, 319-86-8, 60-57-1, 959-98-8, 33213-65-9, 1031-07-8, 72-20-8, 7421-93-4, 53494-70-5, 58-89-9, 76-44-8, 1024-57-3, 72-43-5, 72-54-8, 72-55-9, 50-29-3
  • UN NumberUN1090
  • Shelf Life (months)36
  • DescriptionOrganochlorine Pesticides Mix A, 200 µg/mL in Acetone; 1 mL
Documents & Certificates
Reviews
Q&A