Spex 5252-I Semivolatile Internal Standard for EPA Method 525, 500 µg/mL; 1 mL

Representative image only.

Spex 5252-I Semivolatile Internal Standard for EPA Method 525, 500 µg/mL; 1 mL

Spex | Item: 0455696

Analytical standard for the analysis of drinking water and raw water

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 500 ppm semivolatile internal standard in acetone matrix
  • 1 mL ampule

More About this Item

US EPA Method 525 is an analytical method for the monitoring of semivolatiles in drinking water and raw water sources by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration500 µg/mL (500 ppm)
  • MatrixAcetone
  • Element/IonAcenaphthene-d10, Chrysene-d12, and Phenanthrene-d10
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Acetone; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant
  • UN NumberUN1090
  • CAS Number15067-26-2, 1719-03-5, 1517-22-2
  • Shelf Life (months)36
  • DescriptionSemivolatile Internal Standard for US EPA Method 525, 500 µg/mL; 1 mL
Documents & Certificates
Reviews
Q&A