Spex 5242-F Fortification Solution for US EPA Method 524.3, 1,000 µg/mL; 1 mL

Representative image only.

Spex 5242-F Fortification Solution for US EPA Method 524.3, 1,000 µg/mL; 1 mL

Spex | Item: 0455633

Analytical standards for drinking water

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm fortification solution in methanol-P&T matrix
  • 1 mL ampule

More About this Item

US EPA Method 524.3 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to Methanol 524.3 Revision 4 are supplied in a 24-component mix.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • Element/Ion1,2-Dichlorobenzene-d4; 4-Bromofluorobenzene; Fluorobenzene
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • UN NumberUN1230
  • CAS Number2199-69-1, 460-00-4, 462-06-6
  • Shelf Life (months)36
  • DescriptionFortification Solution for US EPA Method 524.3, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A