Spex 5081-PC Performance Check Solution for US EPA Method 508; 1 mL

Representative image only.

Spex 5081-PC Performance Check Solution for US EPA Method 508; 1 mL

Spex | Item: 0455881

Test for pescides and PBCs in soil, source water, drinking water, and waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 508
  • Multi-component performance check solution in methyl tertiary-butyl ether:ethyl acetate matrix
  • 1 mL ampule

More About this Item

The EPA is tasked with the monitoring of environmental systems as they pertain to contamination and human health. The methods issued by the EPA are created to respond to specific toxins or persistent organic pollutants (POPs) found in environmental samples such as soil, source water, drinking water, and waste. In particular, pollutants such as polychlorinated biphenyls (PCBs), which were in widespread industrial use up until their restriction, are of concern due to their stable and persistent nature in the environment. Another group of chemicals of high concern is the hundreds of commercial pesticides and herbicides in use in the world today. Pesticides, from algaecides to virucides, are used in large quantities in industrial and private agriculture. The concern over human pesticide exposure over the past few decades has led to extensive monitoring of these pesticides. It has been reported that over 98% of insecticides and 95% of herbicides affect areas other than their intended target product. It is essential that monitoring agencies have accurate standard mixes to measure the pesticide levels in the environment.

Many new pesticides are now being tested using highly sensitive LC/MS techniques, in addition to traditional GC techniques, to determine minute amounts of residue in environmental samples and food products. We facilitate ease of monitoring and testing of pesticides by creating pesticide test mixes to suit your monitoring needs. Our standards are designed to work within EPA, AOAC and FDA analytical testing methods using all of the leading analytical techniques: LC, LC/MS, GC, and GC/MS. In addition, custom pesticide mixes can be made to your specifications to create a mix that meets your needs.

Applicable Method:
• US EPA Method 508 is an analytical method for the monitoring of organochlorine pesticides and PCBs in drinking water and ground water by GC/ECD.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component solution contains 4 components.

• Performance Check Solution is a 1 mL ampule which contains 50 ng/mL (50 ppb) of Chlorothalonil and Methyl dacthal, 40 ng/mL (40 ppb) of delta-BHC, and 2 ng/mL (2 ppb) of Chlorpyrifos.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixMethyl Tertiary-Butyl Ether:Ethyl Acetate
  • Element/Iondelta-BHC, Chlorothalonil, Chlorpyrifos, Methyl dacthal
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Flammable liquids, n.o.s. (Methyl tert-butyl ether, Ethyl acetate); Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant
  • CAS Number319-86-8, 1897-45-6, 2921-88-2, 1861-32-1
  • UN NumberUN1993
  • Shelf Life (months)12
  • DescriptionPerformance Check Solution in Methyl Tertiary-Butyl Ether:Ethyl Acetate; 1 mL
Documents & Certificates
Reviews
Q&A