Spex 504-AH-PK EDB/DBCP Analytes Mix (High Level), 2,000 µg/mL; 1 mL; 4/pk

Representative image only.

Spex 504-AH-PK EDB/DBCP Analytes Mix (High Level), 2,000 µg/mL; 1 mL; 4/pk

Spex | Item: 0500889

Analytical standards for drinking water and ground water

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 504 and 8011
  • 2,000 ppm EDB/DBCP analytes mix in methanol-P&T matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 504 is an analytical method for the monitoring of 1,2-Dibromomethane (EDB), 1,2-Dibromo-3-Chloropropane (DBCP), and 1,2,3-Trichloropropane (123TCP) in water by micro-extraction and either GC/ECD or GC/MS techniques.

US EPA Method 8011 is an analytical method for the monitoring of EDB and DBCP in drinking water and ground water by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryOrganics;Standards
  • Element/Ion1-Butanethiol
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number109-79-5
  • UN NumberUN1230
  • Shelf Life (months)36
  • Description1-Butanethiol Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A