Secador F42029-0000 Techni-dome 360 Vacuum Desiccator, 65 Liter Capacity
Representative image only.
Secador F42029-0000 Techni-dome 360 Vacuum Desiccator, 65 Liter Capacity
Secador | Mfr: F42029-0000| Item: 0651450
The largest domed desiccator of its kind -- up to 4 times more volume than other large benchtop units.
£431.20GBP / Each
£517.44GBP (incl VAT)
£431.20GBP / Each
£517.44GBP (incl VAT)
Buy It Together
Current Item
Secador F42029-0000 Techni-dome 360 Vacuum Desiccator, 65 Liter Capacity
Secador F42029-0000 Techni-dome 360 Vacuum Desiccator, 65 Liter Capacity
0651450
£431.20GBP / Each
£517.44GBP (incl VAT)
+
Secador F42040-0000 PTFE Stopcock for 0651450
0651452
£46.22GBP / Each
£55.46GBP (incl VAT)
=
Total Price 477.42 ADD 2 ITEMS TO CART
Product Description
Includes
Polypropylene stopcock with PTFE plug, and two round polypropylene shelves, 14-1/2" and 9" (36.8 cm and 22.9 cm) in diameter.
Key Features
  • Reclaim benchtop space by conveniently nesting the two halves under the bench after use
  • Clear, seamless polycarbonate construction provides all-around visibility, and resistance to chemicals, shattering, and thermal stress
  • Polypropylene stopcock with PTFE plug turns easily allowing a slow, turbulence-free entrance of air, which minimizes disturbance of contents
More About this Item
Desiccator will hold a full vacuum (29" Hg, 737 mm Hg) at room temperature for 24 hours and remain airtight for extended periods of time even if not under vacuum. The gasket, with an application of vacuum grease assures a tight seal down to 0°C. The stopcock accepts 1/4" I.D. (6.4 mm) tubing.
Specifications
  • Chamber Volume (cu ft)2.3
  • Chamber Volume (Liters)65
  • Plate Size (mm)368
  • Exterior Height (InchShort)18
  • Exterior Height (cm)45.7
  • Diameter (InchShort)20
  • Diameter (cm)50
  • MaterialPolycarbonate
  • ColorClear
  • DescriptionSecador Techni-dome 360 Vacuum Desiccator, 65 Liter Capacity