Scienceware F37805-0008 8 liter Churn Sample Splitter
Representative image only.
Scienceware F37805-0008 8 liter Churn Sample Splitter
Scienceware | Mfr: F37805-0008| Item: 9916002
Mix and divide a water sample into subsamples for testing
£637.24GBP / Each
£764.69GBP (incl VAT)
£637.24GBP / Each
£764.69GBP (incl VAT)
Product Description
Key Features
  • Polyethylene vessel with polypropylene spigot
  • Swing handle included for ease of transportation
More About this Item
Use the sample splitter to efficiently composite representative cross-section samples of water-sediment mixtures from rivers and streams. Composite samples are homogenized and then split into representative subsamples using the churn splitter. The spring-loaded spigot drains the subsamples into containers for water quality analysis.
Specifications
  • MaterialPolyethylene, polypropylene
  • Sample Volume8
  • Height (in)11
  • Diameter (in)8.5
  • Description8 liter Churn Sample Splitter