SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/4" ID x 1/2" OD; 25 Ft
Representative image only.
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/4" ID x 1/2" OD; 25 Ft
SANI-TECH | Mfr: ULTRA-C-250-4| Item: 5020257
Ultra-pure silicone tubing with ultra-low extractable is perfect for biopharmaceutical applications
£166.23GBP / Pkg of 1
£199.48GBP (incl VAT)
£166.23GBP / Pkg of 1
£199.48GBP (incl VAT)
Buy It Together
Current Item
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/4" ID x 1/2" OD; 25 Ft
SANI-TECH Ultra-C Sanitary Silicone Tubing, 1/4" ID x 1/2" OD; 25 Ft
5020257
£166.23GBP / Pkg of 1
£199.48GBP (incl VAT)
+ + +
Cole-Parmer Enlarging Check Valve, Natural Polypropylene, 1/8" ID x 1/4" ID; 10/Pk
5014106
£8.75GBP / Pkg of 10
£10.50GBP (incl VAT)
+
Cole-Parmer HDPE In-Line Filter, Polypropylene, Natural, 45-90 Micron, 1/8" ID;  10/Pk
5014129
£10.30GBP / Pkg of 10
£12.36GBP (incl VAT)
+
KD Scientific KDS 100 Legacy 100 Single-Syringe Infusion Pump; 110V
5100510
£1,359.26GBP / Each
£1,631.11GBP (incl VAT)
=
Total Price 1644.73 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Offers excellent flexibility and superior pump life
  • Platinum cured silicone—shows exceptional biocompatibility
  • Low extractables—perfect for maintaining fluid purity
  • Meets the requirements of USP <88> Class VI, and/or USP <87>, and/or ISO 10993-5
  • Laser etched part numbers and lot codes for traceability
  • Produced and double-bagged in an ISO Class 7 certified cleanroom
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection cleaning and sterilization
  • 1/4" ID x 1/2" OD
  • 25 ft per pack
More About this Item
Manufactured from the finest grade of silicone material, Sani-Tech Ultra-C tubing offers the utmost purity in tubing. Extremely low TOCs ensure minimal contamination by extractables, making this perfect for biopharmaceutical application. Laser etched part numbers and lot codes provide product traceability.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Tubing FormulationSani-Tech® Ultra-C
  • Tubing OD (in)1/2
  • Tubing OD (mm)12.7
  • Tubing ID (in)1/4
  • Tubing ID (mm)6.4
  • Wall Thickness (in)1/8
  • Wall Thickness (mm)3.15
  • Clarity (opaque)Yes
  • Burst Pressure (PSI)53
  • Burst Pressure (bar)3.6
  • Min Temperature (° F)-80
  • Max Temperature (° F)500
  • Min Temperature (° C)-62
  • Max Temperature (° C)260
  • Length (ft)25
  • Sterilization MethodEtO;Gamma;Autoclave
  • Regulatory ApprovalsADCF;European Pharmacopoeia;RoHS;USP
  • Durometer50 Shore A
  • DescriptionUltra-C Sanitary Silicone Tubing, 1/4" ID x 1/2" OD; 25 Ft