Saint Gobain Pure-Fit TruValve Systems, Nylon, 5/Pk
Representative image only.
Saint Gobain Pure-Fit TruValve Systems, Nylon, 5/Pk
Saint Gobain | Mfr: TVC75| Item: 4852422
Reliable, calibrated flow control and tamper-proof closure for critical bioprocess operations.
£221.78GBP / Pkg of 5
£266.14GBP (incl VAT)
£221.78GBP / Pkg of 5
£266.14GBP (incl VAT)
Product Description
Key Features
  • Installs easily over flexible tubing or completed assemblies
  • Provides total flow variability with a calibrated, repeatable closure system
  • Lightweight, reinforced nylon construction prevents flow path imbalance
  • Tamper-proof lockout ensures SOP compliance and secure closure
  • Suitable for all flexible tubing materials up to 0.700" OD
  • Fully autoclavable and gamma-sterilizable for aseptic environments
  • Performs reliably across a wide temperature range
  • Ideal for metering, sampling, SIP isolation, and controlled fluid transfer
More About this Item
The Pure-Fit® Tru Valve is engineered for bioprocessing environments where precision, sterility, and ease of use are non-negotiable. Designed to install directly over standard flexible tubing—or even a finished assembly—the Tru Valve eliminates the need to cut or rebuild lines during set-up. Its lightweight borosilicate-reinforced nylon construction ensures durability without adding stress or imbalance to tubing runs.
With its calibrated closure system, operators gain accurate, repeatable flow control supported by scale markings on both sides of the valve—ideal for validation activities and documented process consistency. Beyond metering fluid flow, the Tru Valve provides a secure, tamper-proof lockout, enabling teams to confidently seal tubing during critical steps such as steam-in-place (SIP) cycles.
Fully autoclavable and compatible with gamma sterilization, the Tru Valve integrates seamlessly into aseptic workflows. It supports all flexible tubing materials up to 0.700" OD, making it an adaptable solution for a wide range of biopharmaceutical processes, including drug development, media transfer, sampling, and bioreactor support systems.
Whether you need controlled flow, validated positioning, or reliable system closure, the Pure-Fit® Tru Valve delivers exceptional performance with simple, operator-friendly functionality.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Pinch Valve Tube Size1/8"ID-5/8"OD
  • Housing MaterialBorosilicate reinforced nylon
  • Max Flow Rate For Water (mL/min)3030
  • Max Pressure (PSI)35
  • Tubing OD (in)0.625
  • DescriptionPure-Fit Tru Valve TV-C-75 reinforced nylon pinch valve for flexible tubing assemblies; pack of 5
  • WarrantyManufacturer defect