Reagents Xylenes, ACS Reagent, Amber Glass Bottle, 500 mL

Representative image only.

Reagents Xylenes, ACS Reagent, Amber Glass Bottle, 500 mL

Reagents | Mfr: C1348000-500C| Item: 6228332

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98.5%
  • Xylene isomers
  • Colorless liquid
  • Density: 0.87
  • Boiling point: 138.3 to 141.4 °C (280.94 to 286.52 °F)
  • Melting poing: -47.9 to -13.3 °C (-54.22 to 8.06 °F)
  • Flash point: 25 °C (77 °F)
  • Refractive index: 1.494 to 1.498nD
  • Solubility information: insoluble
  • 500 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC6H4(CH3)2
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDimethylbenzenes, Xylol
  • Volume (mL)500
  • PackagingAmber glass bottle
  • DOT InformationXYLENES, 3, P.G. III
  • Storage SymbolAmbient
  • CAS Number1330-20-7
  • UN NumberUN1307
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionXylenes, ACS Reagent, Amber Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A