Reagents Urea, Shotted Pellets, Wide-Mouth Container, 500 g

Representative image only.

Reagents Urea, Shotted Pellets, Wide-Mouth Container, 500 g

Reagents | Mfr: C2345500-500A5| Item: 6228615

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White shotted pellets
  • Density: 1.34
  • Boiling point: 135 °C (275 °F)
  • Melting point: 133 °C (271.4 °F), decomposes
  • Solubility information: 590 g/L at 20 °C (68 °F)
  • 500 g wide-mouth container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Linear FormulaNH2CONH2
  • ClassSalts
  • ChemistryOrganics
  • FormSolid
  • Weight (g)500
  • Formula Weight60.06
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number57-13-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionUrea, Shotted Pellets, Wide-Mouth Container, 500 g
Documents & Certificates
Reviews
Q&A