Reagents Triphenyltetrazolium Chloride, Purified, Wide-Mouth Amber Glass Bottle, 5 g

Representative image only.

Reagents Triphenyltetrazolium Chloride, Purified, Wide-Mouth Amber Glass Bottle, 5 g

Reagents | Mfr: C2342700-5C2| Item: 6228612

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98%
  • White to yellow solid
  • 5 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaC19H15ClN4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Synonyms2,3,5-Triphenyltetrazolium chloride
  • Weight (g)5
  • Formula Weight334.8
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number298-96-4
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionTriphenyltetrazolium Chloride, Purified, Wide-Mouth Amber Glass Bottle, 5 g
Documents & Certificates
Reviews
Q&A