Reagents Trifluoroacetic Acid, Amber Glass Bottle, 500 mL

Representative image only.

Reagents Trifluoroacetic Acid, Amber Glass Bottle, 500 mL

Reagents | Mfr: C2342660-500X| Item: 6228610

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless to faint yellow liquid
  • Density: 1.54
  • Odor: pungent
  • Solubility information: soluble
  • pH: <1
  • 500 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaCF3COOH
  • ClassAcid
  • FormLiquid
  • Volume (mL)500
  • Formula Weight114.02
  • PackagingAmber glass bottle
  • DOT InformationTRIFLUOROACETIC ACID, 8, P.G. I
  • Storage SymbolAmbient
  • UN NumberUN2699
  • CAS Number76-05-1
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)72.4
  • Boiling Point F (° F)162.32
  • Melting Point C (° C)-15.4
  • Melting Point F (° F)4.28
  • DescriptionTrifluoroacetic Acid, Amber Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A