Reagents Trichloroethylene, ACS Reagent, Amber Glass Bottle, 500 mL

Representative image only.

Reagents Trichloroethylene, ACS Reagent, Amber Glass Bottle, 500 mL

Reagents | Mfr: C1342000-500C| Item: 6228331

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.5%
  • Colorless liquid
  • Density: 1.47
  • Flash point: 90 °C (194 °F)
  • Refractive index: 1.476 to 1.478nD
  • Solubility information: 0.2 g/L at 25 °C (77 °F)
  • 500 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC2HCl3
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)500
  • Formula Weight131.39
  • Vapor Pressure (torr)64.505
  • PackagingAmber glass bottle
  • DOT InformationTRICHLOROETHYLENE, 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN1710
  • CAS Number79-01-6
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)86.7
  • Boiling Point F (° F)188.06
  • Melting Point C (° C)-84.8
  • Melting Point F (° F)-120.64
  • DescriptionTrichloroethylene, ACS Reagent, Amber Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A