Reagents Tributyl Phosphate, Purified, Drum, 200 L

Representative image only.

Reagents Tributyl Phosphate, Purified, Drum, 200 L

Reagents | Mfr: C2341500-200E| Item: 6228609

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless to slightly yellow liquid
  • Density: 0.98
  • Boiling point: 289 °C (552.2 °F), decomposes
  • Flash point: 146 °C (294.8 °F) open cup
  • Refractive index: 1.423 to 1.425nD
  • Solubility information: 0.28 g/L at 25 °C (77 °F)
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear Formula(CH3(CH2)3O)3PO
  • ClassEster
  • ChemistrySolvents
  • SynonymsTBP, TBPA, Butyl phosphate
  • FormLiquid
  • Volume (mL)200000
  • Formula Weight266.31
  • PackagingDrum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number126-73-8
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)-80
  • Melting Point F (° F)-112
  • DescriptionTributyl Phosphate, Purified, Drum, 200 L
Documents & Certificates
Reviews
Q&A