Reagents Toluene, ACS Reagent, Safety Coated Amber Glass Bottle, 4 L

Representative image only.

Reagents Toluene, ACS Reagent, Safety Coated Amber Glass Bottle, 4 L

Reagents | Mfr: C1341000-4C1| Item: 6228329

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.5%
  • Colorless liquid
  • Density: 0.87
  • Boiling point: approximately 111 °C (231.8 °F)
  • Flash point: 4 °C (39.2 °F)
  • Solubility information: 0.52 g/L at 20 °C (68 °F)
  • 4 L safety coated amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC6H5CH3
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight92.14
  • Vapor Pressure (torr)22.502
  • PackagingSafety coated amber glass bottle
  • DOT InformationTOLUENE, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number108-88-3
  • UN NumberUN1294
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)-95
  • Melting Point F (° F)-139
  • Refractive Index1.496
  • DescriptionToluene, ACS Reagent, Safety Coated Amber Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A