Reagents Sulfur, Sublimed, Natural Poly Container, 3 kg

Representative image only.

Reagents Sulfur, Sublimed, Natural Poly Container, 3 kg

Reagents | Mfr: C2335500-3A5| Item: 6228605

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Sublimed yellow solid
  • Density: 2.06
  • Solubility information: insoluble
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Linear FormulaS
  • ClassSalts
  • ChemistryInorganics
  • Element/IonSulfur
  • FormSolid
  • Weight (g)3000
  • Formula Weight32.07
  • PackagingPoly container
  • DOT InformationSULFUR, 4.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number7704-34-9
  • UN NumberUN1350
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)444.7
  • Boiling Point F (° F)832.46
  • Melting Point C (° C)106.8
  • Melting Point F (° F)224.24
  • DescriptionSulfur, Sublimed, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A