Reagents Starch, Potato, Natural Poly Container, 1 kg

Representative image only.

Reagents Starch, Potato, Natural Poly Container, 1 kg

Reagents | Mfr: C2331500-1KG| Item: 6228604

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White solid
  • Density: 1.5
  • Melting point: 255 to 258 °C (491 to 496.4 °F)
  • Solubility information: insoluble
  • 1 kg natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear Formula(C6H10O5)n
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)1000
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number9005-84-9
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionStarch, Potato, Natural Poly Container, 1 kg
Documents & Certificates
Reviews
Q&A