Reagents Stannous Chloride Dihydrate, ACS Reagent, Natural Poly Container, 2.25 kg

Representative image only.

Reagents Stannous Chloride Dihydrate, ACS Reagent, Natural Poly Container, 2.25 kg

Reagents | Mfr: C1330000-2.5A5| Item: 6228324

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White to off-white solid
  • Density: 2.71
  • Melting point: 37 to 38 °C (98.6 to 100.4 °F), decomposes
  • Solubility information: 1780 g/L at 10 °C (50 °F)
  • 2.5 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS
  • Linear FormulaSnCl2·2H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsTin (II) chloride 2-hydrate
  • Weight (g)2500
  • Formula Weight225.65
  • PackagingPoly container
  • DOT InformationCORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. (TIN CHLORIDE), 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number10025-69-1
  • UN NumberUN3260
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Boiling Point C (° C)652
  • Boiling Point F (° F)1205.6
  • DescriptionStannous Chloride Dihydrate, ACS Reagent, Natural Poly Container, 2.25 kg
Documents & Certificates
Reviews
Q&A