Reagents Sodium Sulfite, Purified, Poly Pail, 10 kg

Representative image only.

Reagents Sodium Sulfite, Purified, Poly Pail, 10 kg

Reagents | Mfr: C2326500-10KG| Item: 6228601

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White solid
  • Density: 2.63
  • Melting point: >500 °C (>932 °F), decomposes
  • Solubility information: 307 g/L at 25 °C (77 °F)
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2SO3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)10000
  • Formula Weight126.04
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7757-83-7
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Sulfite, Purified, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A