Reagents Sodium Sulfide, Fused Flakes, Technical, Fiber Drum, 12 kg

Representative image only.

Reagents Sodium Sulfide, Fused Flakes, Technical, Fiber Drum, 12 kg

Reagents | Mfr: C2326000-12F4| Item: 6228600

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless to slightly yellow fused flakes
  • Density: 1.43
  • Solubility information: 206 g/L at 25 °C (77 °F)
  • 12 kg fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeTechnical
  • Linear FormulaNa2S·9H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)12000
  • Formula Weight240.18
  • PackagingFiber drum
  • DOT InformationSODIUM SULFIDE, HYDRATED, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number27610-45-3
  • UN NumberUN1849
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)950
  • Melting Point F (° F)1742
  • DescriptionSodium Sulfide, Fused Flakes, Technical, Fiber Drum, 12 kg
Documents & Certificates
Reviews
Q&A