Reagents Sodium Silicate, 42° Baumé, Drum, 200 L

Representative image only.

Reagents Sodium Silicate, 42° Baumé, Drum, 200 L

Reagents | Mfr: C2325000-200E| Item: 6228597

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • 42° Baumé
  • Colorless/white liquid
  • Density: 1.39
  • Solubility information: miscible
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • ClassSolution
  • ChemistryAnalytical Reagents
  • FormLiquid
  • Volume (mL)200000
  • PackagingDrum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number1344-09-8
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)102
  • Boiling Point F (° F)215.6
  • DescriptionSodium Silicate, 42° Baumé, Drum, 200 L
Documents & Certificates
Reviews
Q&A