Reagents Sodium Phosphate Tribasic Dodecahydrate, Purified, Poly Pail, 10 kg

Representative image only.

Reagents Sodium Phosphate Tribasic Dodecahydrate, Purified, Poly Pail, 10 kg

Reagents | Mfr: C2324500-10KG| Item: 6228593

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white crystals
  • Density: 1.6
  • Melting point: 75 °C (167 °F), decomposes
  • Solubility information: 144 g/L at 25 °C (77 °F)
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaNa3PO4·12H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsTrisodium phosphate dodecahydrate
  • Weight (g)10000
  • Formula Weight380.12
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10101-89-0
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionSodium Phosphate Tribasic Dodecahydrate, Purified, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A